Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SANT-1
GREENER_CHOICESDP

Catalog No. 197410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-741-0 10 mg
19-745-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-741-0 Supplier Tocris Bioscience™ Supplier No. 1974/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibitor of hedgehog (Hh) signaling; antagonizes smoothened activity

SANT-1 is a potent, cell-permeable inhibitor of Sonic hedgehog (Shh) signaling; high affinity antagonist of smoothened activity (KD = 1.2 nM). Inhibits smoothened agonist effects with an IC50 of 20 nM (in Shh-LIGHT2 cells). Has been used in protocols to generate pancreatic β cells from hPSCs.

For more information about how SANT-1 may be used, see our protocol:

Generation of β cells from hPSCs

Chemical Identifiers

CAS 304909-07-7
Molecular Formula C23H27N5

Specifications

Quantity 10 mg
Formula Weight 373.5
Purity >99%
Molecular Weight (g/mol) 373.50
Chemical Name or Material SANT-1
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.