Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAP130 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001077 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001077 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001077 Supplier Novus Biologicals Supplier No. NB1001077
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 3 publications

SAP130 Polyclonal specifically detects SAP130 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Peptide ELISA, Western Blot.

Specifications

Antigen SAP130
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, ELISA, Immunohistochemistry-Frozen, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q15393
Gene Alias KIAA0017splicing factor 3b, subunit 3, 130kD, Pre-mRNA-splicing factor SF3b 130 kDa subunit, RSE1, SAP130SAP 130, SF3B130, SF3b130pre-mRNA splicing factor SF3b, 130 kDa subunit, Spliceosome-associated protein 130, splicing factor 3B subunit 3, splicing factor 3b, subunit 3, 130kDa, STAF130
Gene Symbols SF3B3
Host Species Goat
Immunogen Peptide with sequence C-VSKKLEDIRTRYAF corresponding to C-Terminus according to NP_036558.3.
Molecular Weight of Antigen 136 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23450
Test Specificity Human. Predicted cross-reactivity based on sequence identity: Mouse, Rat, Dog, Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.