Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAP155 Antibody (6C7), Novus Biologicals™
SDP

Catalog No. NB298324 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298324 50 μL
NB298325 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298324 Supplier Novus Biologicals Supplier No. NBP32632650UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SAP155 Monoclonal antibody specifically detects SAP155 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen SAP155
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 6C7
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias Hsh155, pre-mRNA processing 10, pre-mRNA splicing factor SF3b, 155 kDa subunit, Pre-mRNA-splicing factor SF3b 155 kDa subunit, Prp10, PRPF10, SAP 155, SAP155splicing factor 3b, subunit 1, 155kD, SF3B155, SF3b155PRP10, spliceosome associated protein 155, Spliceosome-associated protein 155, splicing factor 3B subunit 1, splicing factor 3b, subunit 1, 155kDa
Host Species Rabbit
Immunogen A synthesized peptide derived from Human SAP155 [UniProt O75533]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23451
Target Species Human, Mouse
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.