Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAP155 Antibody, Novus Biologicals™
SDP

Catalog No. NB10055255 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10055255 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10055255 Supplier Novus Biologicals Supplier No. NB10055255
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SAP155 Polyclonal specifically detects SAP155 in Human samples. It is validated for Immunoprecipitation, KnockDown, Simple Western, Western Blot.

Specifications

Antigen SAP155
Applications Immunoprecipitation, KnockDown, Simple Western, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Simple Western, Immunoprecipitation 2-5 ug/mg lysate, Knockdown Validated
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. O75533
Gene Alias Hsh155, pre-mRNA processing 10, pre-mRNA splicing factor SF3b, 155 kDa subunit, Pre-mRNA-splicing factor SF3b 155 kDa subunit, Prp10, PRPF10, SAP 155, SAP155splicing factor 3b, subunit 1, 155kD, SF3B155, SF3b155PRP10, spliceosome associated protein 155, Spliceosome-associated protein 155, splicing factor 3B subunit 1, splicing factor 3b, subunit 1, 155kDa
Gene Symbols SF3B1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human splicing factor 3b, subunit 1, 155kDa using the numbering given in entry NP_036565.2 (GeneID 23451).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23451
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.