Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAP30 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB057460 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB057460 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB057460 Supplier Novus Biologicals Supplier No. NBP299031H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SAP30 Polyclonal specifically detects SAP30 in Human samples. It is validated for Western Blot.

Specifications

Antigen SAP30
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias 30 kDa Sin3-associated polypeptide, histone deacetylase complex subunit SAP30, Sin3 corepressor complex subunit SAP30, Sin3A-associated protein, 30kDa, Sin3-associated polypeptide p30, sin3-associated polypeptide, 30 kDa, Sin3-associated polypeptide, 30kDa
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human SAP30.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8819
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.