Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAP30BP Antibody (OTI4H1), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP273987FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273987FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273987FR Supplier Novus Biologicals Supplier No. NBP273987FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SAP30BP Monoclonal specifically detects SAP30BP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SAP30BP
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4H1
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias HCNGPDKFZp586L2022, HTRGHSV-1 binding, HTRPSAP30-binding protein, SAP30 binding protein, Transcriptional regulator protein HCNGP
Gene Symbols SAP30BP
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-260 of human SAP30BP (NP_037392) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 29115.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.