Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAP30L Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB100491 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100491 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100491 Supplier Novus Biologicals Supplier No. NB100491
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SAP30L Polyclonal specifically detects SAP30L in Human samples. It is validated for Western Blot.

Specifications

Antigen SAP30L
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Alias DKFZp667L2214, FLJ11526, FLJ36497, HCV non-structural protein 4A-transactivated protein 2, histone deacetylase complex subunit SAP30L, NS4ATP2FLJ23595, SAP30-like, Sin3 corepressor complex subunit SAP30L, Sin3A associated protein p30-like, Sin3-associated protein p30-like
Gene Symbols SAP30L
Host Species Rabbit
Immunogen A synthetic peptide made to residues 165-183 (VKSNKSRLDQKSEGGKQLE) of human SAP30L.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79685
Test Specificity This is specific for SAP30L.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.