Learn More
Azure Biosystems Sapphire FL Standalone Laser
SDP
Catalog No. IS4023AZ Shop All Azure Biosystems Products
Change view
Click to view available options
Wavelength:
375 nm
488 (YFP) nm
488 nm
532 nm
638 nm
658 nm
685 nm
784 nm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wavelength
IS4023AZ 375 nm
IS4025AZ 488 nm
IS4026AZ 532 nm
IS4055AZ 488 (YFP) nm
IS4059AZ 658 nm
IS4027AZ 638 nm
IS4028AZ 685 nm
IS4029AZ 784 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. IS4023AZ Supplier Azure Biosystems Supplier No. IS4023
Only null left
Add to Cart
Add to Cart
Request a Quote
Standalone laser module for Sapphire FL.
  • Standalone laser module for Sapphire FL
  • Does not include emission filter.
  • Available in a variety of different wavelengths
Specifications
Wavelength 375 nm
Product Type Laser Module