Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One Sapphire PCR Tube without Cap
Shop All Greiner Bio-One Products
Click to view available options
Color:
Natural
Description
- Thin-walled for heat transfer during amplification
- Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
- Packaging labelled with product symbols for warehousing
- Used for sample handling

Specifications
Specifications
Color | Natural |
Volume (Metric) | 0.2 mL |
For Use With (Equipment) | PCR |
Sterility | Non-sterile |
Material | Polypropylene |
Product Type | PCR Tube without Cap |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction