Learn More
Greiner Bio-One Sapphire RT-PCR 8-Tube Strips and Ergonomic Caps Combo
Click to view available options
Closure Type:
Flat Strip Cap
Optical Strip Cap
Description
- Thin-walled for heat transfer during amplification
- Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
- Packaging labelled with product symbols for warehousing
- Used for sample handling
Specifications
Specifications
Volume (Metric) | 0.2 mL |
For Use With (Equipment) | Real-time/quantitative PCR |
Sterility | Non-sterile |
Material | Polypropylene |
Closure Type | Flat Strip Cap |
Product Type | RT-PCR 8-Tube Strips and Caps Combo |