Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAR1B Antibody (1C7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP222566 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222566 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222566 Supplier Novus Biologicals Supplier No. NBP222566
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SAR1B Monoclonal specifically detects SAR1B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SAR1B
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1C7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence
Gene Accession No. NP_057187
Gene Alias 2310075M17Rik, ANDD, CMRD, EC 3.6.5, GTP-binding protein B, GTP-binding protein SAR1b, GTP-binding protein Sara, SAR1 homolog B (S. cerevisiae), SAR1a gene homolog (S. cerevisiae) 2, SAR1a gene homolog 2, SAR1a gene homolog 2 (S. cerevisiae), SARA2GTBPB, SARB
Gene Symbols SAR1B
Host Species Mouse
Immunogen Recombinant human SAR1B (1-198aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51128
Test Specificity Anti-human SAR1B mAb, clone 1C7, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human SAR1B protein.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.