Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sarcalumenin Antibody (XIIC4), PE, Novus Biologicals™
SDP

Catalog No. NB344709 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344709 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344709 Supplier Novus Biologicals Supplier No. NB1202874PE

Mouse Monoclonal Antibody

Sarcalumenin Monoclonal antibody specifically detects Sarcalumenin in Mouse, Canine, Guinea Pig, Rabbit samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen Sarcalumenin
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone XIIC4
Conjugate PE
Dilution Western Blot, Immunofluorescence
Formulation PBS
Gene Alias sarcalumenin
Host Species Mouse
Immunogen Purified rabbit sarcoplasmic reticulum membrane vesicles.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 6345
Target Species Mouse, Canine, Guinea Pig, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.