Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS 3b Antibody, Novus Biologicals™
SDP

Catalog No. NB10056746 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056746 0.2 mL
NB10056746T 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056746 Supplier Novus Biologicals Supplier No. NB10056746
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SARS 3b Polyclonal specifically detects SARS 3b in SARS-CoV samples. It is validated for Western Blot.

Specifications

Antigen SARS 3b
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation Whole antisera with PBS and Glycerol with 0.05% Sodium Azide
Gene Accession No. P59633-1
Gene Alias 3b, 3b protein, hypothetical protein sars3b, Hypothetical protein X2, Orf3b, ORF4
Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 31-45 (C-KVTAFQHQNSKKTTK) from the 3b (Non-structural protein 3b) for the Human SARS coronavirus (Genbank accession no. NP_828853.1.)
Purification Method Unpurified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The NCBI accession number for SARS-3b is NP_828853.1.
Target Species SARS-CoV
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.