Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS 3b Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | SARS 3b |
---|---|
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056746T
![]() |
Novus Biologicals
NB10056746SS |
0.05 mL |
Each for $181.90
|
|
|||||
NB10056746
![]() |
Novus Biologicals
NB10056746 |
0.2 mL |
Each for $402.00
|
|
|||||
Description
SARS 3b Polyclonal specifically detects SARS 3b in SARS-CoV samples. It is validated for Western Blot.Specifications
SARS 3b | |
Western Blot | |
Unconjugated | |
Rabbit | |
SARS-CoV | |
P59633-1 | |
The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 31-45 (C-KVTAFQHQNSKKTTK) from the 3b (Non-structural protein 3b) for the Human SARS coronavirus (Genbank accession no. NP_828853.1.) | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Antisera | |
RUO | |
Whole antisera with PBS and Glycerol with 0.05% Sodium Azide | |
3b, 3b protein, hypothetical protein sars3b, Hypothetical protein X2, Orf3b, ORF4 | |
IgG | |
Unpurified | |
The NCBI accession number for SARS-3b is NP_828853.1. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title