Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Envelope Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB185916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185916 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB185916 Supplier Novus Biologicals Supplier No. NBP307060B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 Envelope Polyclonal antibody specifically detects SARS-CoV-2 Envelope in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence

Specifications

Antigen SARS-CoV-2 Envelope
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation PBS with 0.05% Sodium Azide
Gene Alias 2019-nCoV Envelope protein, COVID-19 Envelope protein, E Protein, Envelope, Human Coronavirus Envelope protein, SARSCoV2, SARS-CoV-2 E protein, SARSCoV2 Envelope protein, SARS-CoV-2 Envelope protein, Severe acute respiratory syndrome 2 Envelope Protein
Host Species Rabbit
Immunogen This antibody was raised against a peptide corresponding to 14 amino acids near the carboxyl terminus of SARS-CoV-2 (COVID-19) Envelope protein. The immunogen is located within the last 50 amino acids of SARS-CoV-2 (COVID-19) Envelope protein.
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740570
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.