Learn More
Description
Specifications
Specifications
| Antigen | SARS-CoV-2 nsp1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA 1:4000-1:8000 |
| Formulation | PBS, pH 7.4, 50% glycerol. |
| Gene Alias | Non-structural protein 1, nsp1, ORF1a polyprotein, ORF1ab polyprotein, pp1a, Replicase polyprotein 1a |
| Host Species | Rabbit |
| Immunogen | Recombinant SARS-CoV-2 NSP1(Met1-Gly180) |
| Purification Method | Antigen Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
