Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 nsp13 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB340990 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340990 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340990 Supplier Novus Biologicals Supplier No. NBP307056MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SARS-CoV-2 nsp13 Polyclonal antibody specifically detects SARS-CoV-2 nsp13 in SARS-CoV-2 samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen SARS-CoV-2 nsp13
Applications ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias helicase, Non-structural protein 13
Host Species Rabbit
Immunogen This antibody was raised against a peptide corresponding to 14 amino acids near the center of the SARS-CoV-2 (COVID-19) NSP13 (Helicase) protein. The immunogen is located between 440-490 amino acids of SARS-CoV-2 (COVID-19) NSP13 (Helicase).
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species SARS-CoV-2
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.