Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 nsp16 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB294958 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294958 Supplier Novus Biologicals Supplier No. NBP325659

Rabbit Polyclonal Antibody

SARS-CoV-2 nsp16 Polyclonal antibody specifically detects SARS-CoV-2 nsp16 in SARS-CoV-2 samples. It is validated for Western Blot

Specifications

Antigen SARS-CoV-2 nsp16
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000
Formulation PBS (pH7), 20% Glycerol
Gene Alias Non-structural protein 16, nsp16, ORF1a polyprotein, ORF1ab polyprotein, pp1a, Replicase polyprotein 1a
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of SARS-CoV-2 nsp16. The exact sequence is proprietary.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species SARS-CoV-2
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.