Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 nsp16 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB151330 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151330 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151330 Supplier Novus Biologicals Supplier No. NBP307965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SARS-CoV-2 nsp16 Polyclonal specifically detects SARS-CoV-2 nsp16 in SARS-CoV-2 samples. It is validated for Western Blot, ELISA.

Specifications

Antigen SARS-CoV-2 nsp16
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:4000-1:8000
Formulation PBS, pH 7.4, 50% glycerol.
Gene Alias Non-structural protein 16, nsp16, ORF1a polyprotein, ORF1ab polyprotein, pp1a, Replicase polyprotein 1a
Host Species Rabbit
Immunogen Recombinant SARS-CoV-2 NSP16(Ser6799-Asn7096)
Purification Method Antigen Affinity-purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species SARS-CoV-2
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.