Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 nsp4 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB185705 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185705 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB185705 Supplier Novus Biologicals Supplier No. NBP307050AF750
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 nsp4 Polyclonal antibody specifically detects SARS-CoV-2 nsp4 in SARS-CoV-2 samples. It is validated for ELISA, Immunofluorescence

Specifications

Antigen SARS-CoV-2 nsp4
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Corona_NSP4_C, Coronavirus nonstructural protein 4 C-terminus, Non-structural protein 4, nsp4
Host Species Rabbit
Immunogen This antibody was raised against a peptide corresponding to 15 amino acids near the center of SARS-CoV-2 (COVID-19) NSP4 protein. The immunogen is located within 50-100 amino acids of the SARS-CoV-2 (COVID-19) NSP4 protein.
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.