Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 nsp9 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB341037 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341037 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341037 Supplier Novus Biologicals Supplier No. NBP307044PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SARS-CoV-2 nsp9 Polyclonal antibody specifically detects SARS-CoV-2 nsp9 in SARS-CoV-2 samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen SARS-CoV-2 nsp9
Applications ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate PerCP
Dilution ELISA, Immunohistochemistry
Formulation PBS
Gene Alias Non-structural protein 9, nsp9 replicase
Host Species Rabbit
Immunogen This antibody was raised against a peptide corresponding to 13 amino acids near the center of SARS-CoV-2 (COVID-19) NSP9 protein. The immunogen is located within 30-80 amino acids of the SARS-CoV-2 (COVID-19) NSP9 protein.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species SARS-CoV-2
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.