Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Nucleocapsid Antibody (019), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB383190 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383190 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383190 Supplier Novus Biologicals Supplier No. NBP312735V
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SARS-CoV-2 Nucleocapsid Monoclonal antibody specifically detects SARS-CoV-2 Nucleocapsid in SARS-CoV samples. It is validated for Western Blot, ELISA

Specifications

Antigen SARS-CoV-2 Nucleocapsid
Applications Western Blot, ELISA
Classification Monoclonal
Clone 019
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias 2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9; structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein
Host Species Rabbit
Immunogen Recombinant SARS-CoV-2 Nucleocapsid Protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 43740575
Target Species SARS-CoV
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.