Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Nucleocapsid Protein Monoclonal Antibody (1C7), Bioss
SDP

Catalog No. 502279698
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9698 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-227-9698 Supplier Bioss Supplier No. BSM41411M
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SARS-CoV-2 Nucleocapsid Protein Monoclonal antibody specifically detects SARS-CoV-2 Nucleocapsid Protein in Virus samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SARS-CoV-2 Nucleocapsid Protein
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1C7
Conjugate Unconjugated
Formulation PBS with 0.1% ProClin 300
Gene N
Gene Accession No. P59595
Gene Alias COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid
Host Species Mouse
Immunogen Recombinant SARS-CoV-2 N protein (His-tag)
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489678
Target Species Virus
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.