Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SARS-CoV-2 Nucleocapsid Recombinant Rabbit Monoclonal Antibody (4Z6T2), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA536286
Description
Endotoxin level <1.0 EU/μg of the protein by LAL method. MA5-36286 has been used in ELISA as a detection antibody with MA5-36284 as a capture antibody.
SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus. The SARS-CoV-2 N protein is an abundant RNA-binding protein critical for viral genome packaging, yet the molecular details that involve this process are poorly understood. N protein contains three dynamic disordered regions that house putative transiently-helical binding motifs. The two folded domains interact minimally such that full-length N protein is a flexible and multivalent RNA-binding protein. SARS-CoV-2 causes illnesses ranging from the common cold to severe diseases.
Specifications
SARS-CoV-2 Nucleocapsid | |
Recombinant Monoclonal | |
1.9 mg/mL | |
PBS with 0.1% Braveds MB-1; pH 7.4 | |
P0DTC9 | |
N | |
HK293 derived SARS-COV-2 Nucleocapsid corresponding to amino acids 2-419. | |
100 μg | |
Primary | |
Virus | |
Antibody | |
IgG |
ELISA, Functional Assay, Surface Plasmon Resonance | |
4Z6T2 | |
Unconjugated | |
N | |
COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N | |
Rabbit | |
Affinity chromatography | |
RUO | |
43740575 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction