Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 ORF3a Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB186172 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB186172 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB186172 Supplier Novus Biologicals Supplier No. NBP311933AF750
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 ORF3a Polyclonal antibody specifically detects SARS-CoV-2 ORF3a in SARS-CoV-2 samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen SARS-CoV-2 ORF3a
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 2019-nCoV ORF3a, 2019-nCoV ORF3a Protein, COVID-19 ORF3a, Human coronavirus ORF3a Protein, ORF3a protein, SARS-CoV-2, SARS-CoV-2 Accessory protein 3a, SARSCoV2 ORF3a Protein, SARS-CoV-2 ORF3a Protein, SARS-CoV-2 Protein 3a, SARS-CoV-2 Protein U274, SARS-CoV-2 Protein X1, Severe Acute Respiratory Syndrome Coronavirus 2
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 15 amino acids near the amino-terminus of SARS-CoV-2 ORF3a protein. The immunogen is located within the first 50 amino acids of the SARS-CoV-2 ORF3a protein.
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740569
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.