Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 ORF7a Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB186232 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB186232 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB186232 Supplier Novus Biologicals Supplier No. NBP311936AF594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 ORF7a Polyclonal antibody specifically detects SARS-CoV-2 ORF7a in SARS-CoV-2 samples. It is validated for ELISA, Immunofluorescence

Specifications

Antigen SARS-CoV-2 ORF7a
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 2019-nCoV ORF7a, 2019-nCoV ORF7a Protein, Accessory Protein 7a, COVID-19 Accessory Protein 7a, COVID-19 ORF7a, COVID-19 Protein U122, COVID-19 Protein X4, Human coronavirus ORF7a Protein, ORF7a Protein, Protein U122, Protein X4, SARS-CoV-2 Accessory Protein 7a, SARS-CoV-2 Protein U122, SARS-CoV-2 Protein X4
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 9 amino acids near the carboxyl terminus of SARS-CoV-2 ORF7a protein. The immunogen is located within the last 50 amino acids of the SARS-CoV-2 ORF7a protein.
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740573
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.