Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 ORF8 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB186213 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB186213 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB186213 Supplier Novus Biologicals Supplier No. NBP311935AF700
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 ORF8 Polyclonal antibody specifically detects SARS-CoV-2 ORF8 in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen SARS-CoV-2 ORF8
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 2019-nCoV ORF8, 2019-nCoV ORF8 Protein, COVID-19 Non-structural protein 8, COVID-19 ns8, COVID-19 ORF8, Human coronavirus ORF8 Protein, Non-structural protein 8, ns8, ORF8 protein, SARS-CoV-2, SARS-CoV-2 Non-structural protein 8, SARS-CoV-2 ns8, SARSCoV2 ORF8 Protein, SARS-CoV-2 ORF8 Protein, Severe Acute Respiratory Syndrome Coronavirus 2 ORF8 Protein
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 13 amino acids near the central domain of SARS-CoV-2 ORF8 protein. The immunogen is located within 30-80 amino acids of the SARS-CoV-2 ORF8 protein.
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740577
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.