Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Spike Antibody (CR3022), DyLight 488 - Chimeric, Novus Biologicals™
SDP

Catalog No. NB025759 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB025759 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB025759 Supplier Novus Biologicals Supplier No. NBP290979G
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SARS-CoV-2 Spike Monoclonal antibody specifically detects SARS-CoV-2 Spike in SARS-CoV-2, SARS-CoV samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen SARS-CoV-2 Spike
Applications ELISA
Classification Monoclonal
Clone CR3022
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, Human coronavirus spike glycoprotein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein
Gene Symbols S
Host Species Rabbit
Immunogen The original monoclonal antibody was generated by sequencing peripheral blood lymphocytes of a patient exposed to the SARS-CoV.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody binds to both SARS-CoV and SARS-CoV-2 with high affinity (PMID: 16796401 & 32065055). It binds the amino acids 318-510 in the S1 domain of the SARS-CoV Spike protein as well as SARS-CoV-2 (COVID-19) Spike protein. The antibody also binds to P462L-substituted S318-510 fragments of the SARS spike protein. The binding epitope is only accessible in the "open" confromation of the spike protein (Joyce et al. 2020). NBP2-90979 cross-reacts with spike protein of Omicron and Delta variants.
Target Species SARS-CoV
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.