Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Spike Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB340718 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340718 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340718 Supplier Novus Biologicals Supplier No. NBP312852PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SARS-CoV-2 Spike Polyclonal antibody specifically detects SARS-CoV-2 Spike in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen SARS-CoV-2 Spike
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein
Host Species Rabbit
Immunogen Anti-SARS-CoV-2 Spike 156-157EF antibody was raised against a peptide corresponding to 13 amino acids near the amino terminal domain of SARS-CoV-2 Spike protein. The immunogen is located within 130-180 amino acids of SARS-CoV-2 Spike protein.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 43740568
Target Species SARS-CoV-2
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.