Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Spike Llama anti-SARS-CoV, SARS-CoV-2, Clone: T3P3F4, Novus Biologicals™
SDP

Catalog No. NB175689 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175689 0.1 mg
NB175688 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB175689 Supplier Novus Biologicals Supplier No. NBP3185270.1MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Llama Monoclonal Antibody

SARS-CoV-2 Spike Monoclonal antibody specifically detects SARS-CoV-2 Spike in SARS-CoV, SARS-CoV-2 samples. It is validated for ELISA

Specifications

Antigen SARS-CoV-2 Spike
Applications ELISA
Classification Monoclonal
Clone T3P3F4
Conjugate Unconjugated
Formulation PBS
Gene Alias 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein
Host Species Llama
Immunogen SARS-CoV-2 variant Spike proteins
Purification Method Affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 43740568
Target Species SARS-CoV, SARS-CoV-2
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.