Learn More
SARS-CoV-2 Spike S1 Chimeric Recombinant Human Monoclonal Antibody (hIgA2001), GenScript™
Human Recombinant Monoclonal Antibody
$437.58
Specifications
Antigen | SARS-CoV-2 Spike S1 Chimeric |
---|---|
Clone | hIgA2001 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-212-0773
![]() |
Genscript Corporation
A02071100 |
100 μg |
Each of 1 for $437.58
|
|
|||||
Description
SARS-CoV-2 (Severe acute respiratory syndrome coronavirus 2), also known as 2019-nCoV, is a positive-sense single-stranded RNA virus. It caused coronavirus disease 2019 (COVID-19). SARS-CoV-2 contains glycosylated spike (S) protein, which is composed of S1 subunit and S2 subunit. The S1 contains a receptor-binding domain (RBD) that can bind to ACE2 receptor on target cells.SARS-CoV-2 Spike S1 Antibody (hIgA2001), Human Chimeric is produced from cell culture in vitro under conditions free from animal-derived components.
SARS-CoV-2 Spike S1 Chimeric Recombinant Monoclonal antibody specifically detects SARS-CoV-2 Spike S1 Chimeric in Virus samples. It is validated for ELISASpecifications
SARS-CoV-2 Spike S1 Chimeric | |
1 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with 0.02% sodium azide; pH 7.4 | |
Recombinant SARS-CoV-2 RBD Protein | |
Primary | |
-20°C |
hIgA2001 | |
ELISA | |
Unconjugated | |
Human | |
Virus | |
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
IgA | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.