Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Spike S1 Protein Antibody (1035226), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB175892 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175892 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175892 Supplier Novus Biologicals Supplier No. FAB105805X
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SARS-CoV-2 Spike S1 Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike S1 Protein in SARS-CoV-2 samples. It is validated for Flow Cytometry

Specifications

Antigen SARS-CoV-2 Spike S1 Protein
Applications Flow Cytometry
Classification Monoclonal
Clone 1035226
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias SARS-CoV-2
Host Species Mouse
Immunogen HEK293-derived SARS-CoV-2 Spike S1 Subunit, Accession # YP_009724390.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.