Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Spike S2 Subunit Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB11370100
This item is not returnable.
View return policy
Description
SARS-CoV-2 Spike S2 Monoclonal antibody specifically detects SARS-CoV-2 Spike S2 in SARS-CoV-2 samples. It is validated for Western Blot, ELISASpecifications
SARS-CoV-2 Spike S2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
43740568 | |
SARS-CoV-2 | |
Purified |
Western Blot, ELISA | |
2812A | |
Western Blot 1 ug/mL, ELISA | |
COVID-19, S2 protein, SARS-CoV-2, SARS-CoV-2 Spike, Spike glycoprotein Subunit 2, spike protein, Spike protein S2, structural protein, surface glycoprotein | |
E. coli-derived recombinant human SARS-CoV-2 Spike S2 Subunit, Met697-Pro1213 | |
100 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction