Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SARS-CoV-2 Spike S2 Subunit Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$177.10 - $433.50
Specifications
Antigen | SARS-CoV-2 Spike S2 |
---|---|
Clone | 2812A |
Dilution | Western Blot 1 ug/mL, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11370100
![]() |
R&D Systems
MAB11370100 |
100 μg |
Each for $433.50
|
|
|||||
MAB11370SP
![]() |
R&D Systems
MAB11370SP |
25 μg |
Each for $177.10
|
|
|||||
Description
SARS-CoV-2 Spike S2 Monoclonal antibody specifically detects SARS-CoV-2 Spike S2 in SARS-CoV-2 samples. It is validated for Western Blot, ELISASpecifications
SARS-CoV-2 Spike S2 | |
Western Blot 1 ug/mL, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
43740568 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
2812A | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
SARS-CoV-2 | |
COVID-19, S2 protein, SARS-CoV-2, SARS-CoV-2 Spike, Spike glycoprotein Subunit 2, spike protein, Spike protein S2, structural protein, surface glycoprotein | |
E. coli-derived recombinant human SARS-CoV-2 Spike S2 Subunit, Met697-Pro1213 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title