Learn More
Description
Specifications
Specifications
| Antigen | Sars E2 |
| Applications | Western Blot, ELISA, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 4A6C9 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry-Paraffin |
| Formulation | Subclonal supernatant with No Preservative |
| Host Species | Mouse |
| Immunogen | Purified recombinant fragment of SARS-E2 glycoprotein precursor expressed in E. Coli. |
| Purification Method | Tissue culture supernatant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
