Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sars Envelope Protein Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB355456 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355456 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355456 Supplier Novus Biologicals Supplier No. NBP241062MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sars Envelope Protein Polyclonal antibody specifically detects Sars Envelope Protein in SARS-CoV samples. It is validated for ELISA

Specifications

Antigen Sars Envelope Protein
Applications ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias SARS coronavirus Envelope protein, SARS CoV E protein, SARS CoV Envelope protein, SARSCoV, SARSCoV Envelope protein, Severe acute respiratory syndrome
Host Species Rabbit
Immunogen Anti-Sars Envelope Protein antibody was raised against a peptide corresponding to 11 amino acids near the carboxy terminus of SARS-CoV Envelope protein. The immunogen is located within the last 50 amino acids of SARS Envelope Proteins.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 1489671
Target Species SARS-CoV
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.