Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sars Membrane Protein Antibody (2H2C4), Novus Biologicals™
SDP

Catalog No. NBP12885201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP12885201 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP12885201 Supplier Novus Biologicals Supplier No. NBP1288520.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sars Membrane Protein Monoclonal specifically detects Sars Membrane Protein in SARS-CoV samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Sars M
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2H2C4
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation Subclonal supernatant with No Preservative
Host Species Mouse
Immunogen Purified recombinant fragment of SARS-m protein expressed in E. Coli.
Purification Method Tissue culture supernatant
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Specific for SARS-M.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.