Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sars Membrane Protein Antibody, Novus Biologicals™
SDP

Catalog No. NB10056569 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056569 0.1 mg
NB10056569T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056569 Supplier Novus Biologicals Supplier No. NB10056569
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Sars Membrane Protein Polyclonal specifically detects Sars Membrane Protein in SARS-CoV samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen Sars Membrane Protein
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Immunocytochemistry/Immunofluorescence reported by customer review
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. P59596
Gene Alias E1 glycoprotein, Matrix glycoprotein, Membrane glycoprotein, SARS coronavirus M protein, SARS coronavirus Membrane protein, SARS CoV, SARS CoV M glycoprotein, SARS CoV Matrix protein, Sars M, Sars Matrix Protein, SARSCoV, SARS-CoV M protein, SARS-CoV Membrane protein, Severe acute respiratory syndrome
Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 195-210 (YSRYRIGNYKLNTDH) from the M (Membrane protein) for the Human SARS coronavirus (Genbank accession no. YP_009724393.1)
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489672
Test Specificity The NCBI accession number of the SARS matrix protein is NP_828855.1.
Target Species SARS-CoV
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.