Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sars Membrane Protein Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB220013 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB220013 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB220013 Supplier Novus Biologicals Supplier No. NBP241060H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sars Membrane Protein Polyclonal antibody specifically detects Sars Membrane Protein in SARS-CoV samples. It is validated for Western Blot, ELISA

Specifications

Antigen Sars Membrane Protein
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias E1 glycoprotein, Matrix glycoprotein, Membrane glycoprotein, SARS coronavirus M protein, SARS coronavirus Membrane protein, SARS CoV, SARS CoV M glycoprotein, SARS CoV Matrix protein, Sars M, Sars Matrix Protein, SARSCoV, SARS-CoV M protein, SARS-CoV Membrane protein, Severe acute respiratory syndrome
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 15 amino acids at the carboxy terminus of the SARS matrix protein. The immunogen is located within the last 50 amino acids of SARS Matrix. Amino Acid Squence: GNYKLNTDHAGSNDN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 1489672
Target Species SARS-CoV
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.