Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Nucleocapsid Protein Antibody (001), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB041450 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB041450 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB041450 Supplier Novus Biologicals Supplier No. NBP290988AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SARS Nucleocapsid Protein Monoclonal specifically detects SARS Nucleocapsid Protein in SARS-CoV-2, SARS-CoV samples. It is validated for Western Blot, ELISA.

Specifications

Antigen SARS Nucleocapsid Protein
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias N, N protein, N structural protein, NC, Nucleocapsid protein, Nucleoprotein, Protein N, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARS Nucleoprotein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, Severe acute respiratory syndrome
Gene Symbols N
Host Species Rabbit
Immunogen Recombinant SARS-CoV Nucleoprotein/NP Protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489678
Test Specificity Has cross-reactivity in ELISA and WB with: SARS-CoV-2 Nucleocapsid Protein Has cross-reactivity in ELISA with: SARS-CoV-2 Omicron (B.1.1.529) Nucleocapsid Protein SARS-CoV-2 Nucleocapsid Protein SARS-CoV-2 Alpha(B.1.1.7) Nucleocapsid(R203K, G204R) Protein SARS-CoV-2 Alpha(B.1.1.7) Nucleocapsid(I292T) Protein SARS-CoV-2 Alpha(B.1.1.7) Nucleocapsid(D3L, R203K, G204R, S235F) Protein SARS-CoV-2 Alpha(B.1.1.7) Nucleocapsid(D3L, S235F) Protein SARS-CoV-2 Alpha(B.1.1.7/ B.1.237) Nucleocapsid(S194L) Protein SARS-CoV-2 Alpha/beta(B.1.1.7/ B.1.351/ A.2.2) Nucleocapsid(P13L) Protein SARS-CoV-2 Beta(B.1.351/B.1.351.2/b.1.351.3/B.1.427/B.1.429) Nucleocapsid (T205I) Protein SARS-CoV-2 Gamma(P.1/ P.1.1/ P.1.2) Nucleocapsid (P80R) Protein SARS-CoV-2 Eta(B.1.525) Nucleocapsid (A12G, T205I) Protein SARS-CoV-2 (B.1.617) Nucleocapsid (D377Y) Protein SARS-CoV-2 (B.1.617) Nucleocapsid (R203M, D377Y) Protein SARS-CoV-2 Delta(B.1.617.2) Nucleocapsid (D63G, R203M, D377Y) Protein SARS-CoV-2 (B.1.617.3) Nucleocapsid (P67S, R203M, D377Y) Protein No cross-reactivity in ELISA with: MERS-CoV Nucleocapsid protein HCoV-229E Nucleocapsid protein HCoV-NL63 Nucleocapsid protein HCoV-HKU1(isolate N5) Nucleocapsid protein HCoV-OC43 Nucleocapsid protein
Target Species SARS-CoV-2, SARS-CoV
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.