Learn More
Description
Specifications
Specifications
| Antigen | SARS Nucleocapsid Protein |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1 |
| Conjugate | FITC |
| Dilution | Western Blot, ELISA |
| Formulation | PBS |
| Gene Alias | N, N protein, N structural protein, NC, Nucleocapsid protein, Nucleoprotein, Protein N, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARS Nucleoprotein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, Severe acute respiratory syndrome |
| Gene Symbols | N |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
