Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Nucleocapsid Protein Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB339536 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339536 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339536 Supplier Novus Biologicals Supplier No. NB10056576APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SARS Nucleocapsid Protein Polyclonal specifically detects SARS Nucleocapsid Protein in SARS-CoV, SARS-CoV-2, Virus samples. It is validated for Dual RNAScope ISH-IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), SDS-Page, Western Blot.

Specifications

Antigen SARS Nucleocapsid Protein
Applications Dual RNAScope ISH-IHC, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), SDS-Page, Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, SDS-Page, Dual RNAscope ISH-IHC
Formulation PBS
Gene Alias N, N protein, N structural protein, NC, Nucleocapsid protein, Nucleoprotein, Protein N, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARS Nucleoprotein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, Severe acute respiratory syndrome
Host Species Rabbit
Immunogen The antibody was developed by immunizing Rabbit with a synthetic peptide corresponding to amino acids 399-411 (DLDDFSKQLQQSM-C) from the N (SARS Nucleocapsid) for the Human SARS coronavirus (Genbank accession no. YP_009724397.2)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 1489678
Target Species SARS-CoV, SARS-CoV-2, Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.