Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Nucleocapsid Protein Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB100560501 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100560501 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100560501 Supplier Novus Biologicals Supplier No. NB100560500.1MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SARS Nucleocapsid Protein Polyclonal specifically detects SARS Nucleocapsid Protein in SARS-CoV samples. It is validated for ELISA, Western Blot.

Specifications

Antigen SARS Nucleocapsid Protein
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Formulation Whole antisera with PBS and 0.2% Gelatin with 0.05% Sodium Azide
Gene Alias N, N protein, N structural protein, NC, Nucleocapsid protein, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, Severe acute respiratory syndrome
Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 20-34 of putative SARS nucleocapsid (Genbank accession no. NP_828858).
Purification Method Unpurified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The was tested on a cell line transfected with full-length SARS Nucleocapsid cDNA with a predicted molecular weight of 46 kDa.
Target Species SARS-CoV
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.