Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Nucleocapsid Protein Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10056576 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056576 0.1 mg
NB10056576T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056576 Supplier Novus Biologicals Supplier No. NB10056576
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 49 publications

SARS Nucleocapsid Protein Polyclonal specifically detects SARS Nucleocapsid Protein in Hamster, Human, SARS-CoV, SARS-CoV-2, Virus samples. It is validated for Dual RNAScope ISH-IHC, ELISA, IF/ICC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In Situ Hybridization, Mass Spectrometry, RNAscope compatible, SDS-Page, Western Blot.

Specifications

Antigen SARS Nucleocapsid Protein
Applications Dual RNAScope ISH-IHC, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), Mass Spectrometry, Dual RNAScope ISH-IHC, SDS-Page, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry reported in scientific literature (PMID 32396922), Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Mossel et al (2008)), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24725942), SDS-Page reported in scientific literature (PMID 34880383), Dual RNAscope ISH-IHC
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_006504.2
Gene Alias 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, N, N protein, N structural protein, NC, Nucleocapsid protein, Nucleoprotein, Protein N, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARS Nucleoprotein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, SARS-CoV-2 N protein, Severe acute respiratory syndrome
Host Species Rabbit
Immunogen The antibody was developed by immunizing Rabbit with a synthetic peptide corresponding to amino acids 399-411 (DLDDFSKQLQQSM-C) from the N (SARS Nucleocapsid) for the Human SARS coronavirus (Genbank accession no. YP_009724397.2)
Purification Method Immunogen affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity SARS Nucleocapsid protein. Dot Blot results using recombinant proteins for cross-reactivity testing revealed high reactivity to SARS-CoV-2 Nucleocapsid protein (NBP2-90975) and low/no reactivity towards H1N1 (NBP1-99041).
Target Species Hamster, SARS-CoV, Human, SARS-CoV, SARS-CoV-2, Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.