Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Nucleocapsid Protein Antibody, Novus Biologicals™
SDP

Catalog No. NB396391 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396391 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB396391 Supplier Novus Biologicals Supplier No. NBP244205100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SARS Nucleocapsid Protein Polyclonal antibody specifically detects SARS Nucleocapsid Protein in SARS-CoV samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SARS Nucleocapsid Protein
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, ELISA 1:10000 - 1:50000, Immunohistochemistry 1:100 - 1:6000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Gene Alias N, N protein, N structural protein, NC, Nucleocapsid protein, Nucleoprotein, Protein N, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARS Nucleoprotein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, Severe acute respiratory syndrome
Host Species Rabbit
Immunogen This SARS Nucleocapsid Protein Antibody was developed by immunizing rabbits with a full-length recombinant protein from the N (SARS Nucleocapsid) for the Human SARS coronavirus (Uniprot#: P59595)
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 1489678
Reconstitution For the 500 μg size, we recommend reconstituting with 100 μg deionized water. The 0.1 mg size is provided in liquid format (0.02 mL).
Target Species SARS-CoV
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.