Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS RDRP Antibody (4E6), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB203141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB203141 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB203141 Supplier Novus Biologicals Supplier No. NBP250258JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SARS RDRP Monoclonal antibody specifically detects SARS RDRP in SARS-CoV samples. It is validated for Western Blot

Specifications

Antigen SARS RDRP
Applications Western Blot
Classification Monoclonal
Clone 4000000
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen The antibody was developed by immunizing mice with a GST fusion protein corresponding to Replicase polyprotein 1ab for the Human SARS coronavirus (Genbank accession no. NP_828849.2)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species SARS-CoV
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.