Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Spike Protein Antibody, Novus Biologicals™
SDP

Catalog No. NB10056048T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056048T 0.025 mg
NB10056048 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056048T Supplier Novus Biologicals Supplier No. NB10056048SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

Sars Spike Protein Polyclonal specifically detects Sars Spike Protein in SARS-CoV samples. It is validated for ELISA, In vitro Assay, Western Blot.

Specifications

Antigen Sars Spike Protein
Applications ELISA, In vitro Assay, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1-1 ug/ml, ELISA 0.1-1 ug/ml, In vitro assay reported in scientific literature (PMID 34120764)
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Alias E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, SARS coronavirus S protein, SARS coronavirus Spike Protein, SARS CoV, SARS CoV S glycoprotein, SARS CoV Spike protein, SARSCoV, SARS-CoV S protein, SARS-COV Spike, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2
Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 288-303 (AVDCALDPLSETKCTL) from the S (Spike glycoprotein)(Spike protein 1) for the Human SARS coronavirus (Genbank accession no. YP_009724390)
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489668
Test Specificity The antibody was tested on a cell line transfected with a partial-length cDNA coding for the N-terminal portion of SARS Spike protein with a predicted molecular weight of 68 kDa.
Target Species SARS-CoV
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.