Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Spike Protein Antibody (16F1071), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP224746V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224746V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224746V Supplier Novus Biologicals Supplier No. NBP224746V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SARS Spike Protein Monoclonal specifically detects SARS Spike Protein in SARS-CoV samples. It is validated for Western Blot.

Specifications

Antigen Sars Spike Protein
Applications Western Blot
Classification Monoclonal
Clone 16F1071
Conjugate DyLight 405
Dilution Western Blot
Gene Accession No. P49591
Gene Alias E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2
Host Species Mouse
Immunogen The antibody was developed using a synthetic peptide from the putative SARS Spike glycoprotein (Genbank accession no. NP_828851) corresponding to amino acids 19-45.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 1489668
Test Specificity The C1.18.4 hybridoma cell line was grown and tissue culture supernatant was purified using affinity chromatography.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.