Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Spike Protein Antibody (D005), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB025805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB025805 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB025805 Supplier Novus Biologicals Supplier No. NBP290989C
Add to Cart
Add to Cart

Human Monoclonal Antibody

SARS Spike Protein Monoclonal specifically detects SARS Spike Protein in SARS-CoV-2, SARS-CoV samples. It is validated for ELISA.

Specifications

Antigen Sars Spike Protein
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone D005
Conjugate DyLight 650
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, SARS coronavirus S protein, SARS coronavirus Spike Protein, SARS CoV, SARS CoV S glycoprotein, SARS CoV Spike protein, SARSCoV, SARS-CoV S protein, SARS-COV Spike, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2
Host Species Human
Immunogen Recombinant SARS-CoV Spike RBD Protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Has cross-reactivity in ELISA with: SARS-CoV-2 Omicron (B.1.1.529) Spike RBD Protein SARS-CoV-2 Omicron (B.1.1.529) Spike S1 Protein SARS-CoV-2 Omicron (B.1.1.529) S1+S2 trimer Protein SARS-CoV-2 Delta (B.1.617.2) Spike RBD Protein SARS-CoV-2 Delta (B.1.617.2) Spike S1+S2 Protein SARS-CoV Spike S1 Protein SARS-CoV-2 Spike S1 Protein SARS-CoV-2 Spike RBD Protein No cross-reactivity in ELISA with: MERS-CoV Spike S1 Protein HCoV-HKU1 (isolate N1) Spike S1 Protein HCoV-HKU1 (isolate N5) Spike S1 Protein HCoV-NL63 Spike S1 Protein HCoV-229E Spike S1 Protein HCoV-OC43 Spike S1+S2 ECD Protein
Target Species SARS-CoV-2, SARS-CoV
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.