Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS Spike Protein Antibody (D005), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB315785 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB315785 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB315785 Supplier Novus Biologicals Supplier No. NBP290989MFV500
Add to Cart
Add to Cart

Human Monoclonal Antibody

SARS Spike Protein Monoclonal antibody specifically detects SARS Spike Protein in SARS-CoV, SARS-CoV-2 samples. It is validated for ELISA

Specifications

Antigen SARS Spike Protein
Applications ELISA
Classification Monoclonal
Clone D005
Conjugate mFluor Violet 500 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, SARS coronavirus S protein, SARS coronavirus Spike Protein, SARS CoV, SARS CoV S glycoprotein, SARS CoV Spike protein, SARSCoV, SARS-CoV S protein, SARS-COV Spike, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2
Host Species Human
Immunogen Recombinant SARS-CoV Spike RBD Protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Immunology, Stem Cell Markers, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species SARS-CoV, SARS-CoV-2
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.